Learn More
NUAK2 Polyclonal Antibody

Description
NUAK2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | NUAK2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.61 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Nuak2 |
| Gene Accession No. | Q66HE5, Q9H093 |
| Gene Alias | 1200013B22Rik; EMBL:AAH81899.1}; mKIAA0537; NUAK family kinase 2; NUAK family SNF1-like kinase 2; NUAK family, SNF1-like kinase, 2; NUAK2; nuak2 {ECO:0000312; Omphalocele kinase 2; Omphk2; SNARK; SNF1/AMP activated protein kinase; SNF1/AMP kinase-related kinase; SNF1/AMP-activated protein kinase; UV126 |
| Gene Symbols | Nuak2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.