Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUB1/NYREN18 Antibody (OTI4H2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273084JF646
Description
NUB1/NYREN18 Monoclonal specifically detects NUB1/NYREN18 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NUB1/NYREN18 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NUB1 | |
Recombinant protein fragment corresponding to amino acids 1-326 of human NUB1/NYREN18 (NP_057202) produced in E.coli. | |
0.1 mL | |
51667.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4H2 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
BS4, NEDD8 ultimate buster-1, negative regulator of ubiquitin-like proteins 1NUB1L, NY-REN-18 antigen, NYREN18NEDD8 ultimate buster 1, Renal carcinoma antigen NY-REN-18 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction