Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559736 Supplier Invitrogen™ Supplier No. PA559736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSGFICPKCK KESQIFPPTT GGAELMCQDL EVPLLGRVPL DPLIGKNCDK GQSFFIDAPD SPATLAYRSI IQRIQEFCNL HQSKEENLIS S Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

Component of the cytosolic iron-sulfur (Fe/S) protein assembly (CIA) machinery (PubMed:18573874). Required for maturation of extramitochondrial Fe-S proteins (PubMed:18573874). The NUBP1-NUBP2 heterotetramer forms a Fe-S scaffold complex, mediating the de novo assembly of an Fe-S cluster and its transfer to target apoproteins (PubMed:18573874). Implicated in the regulation of centrosome duplication. Negatively regulates cilium formation and structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NUBP1
Gene Accession No. P53384
Gene Alias cytosolic Fe-S cluster assembly factor NUBP1; cytosolic Fe-S cluster assembly factor NUBP1 {ECO:0000255; EMBL:AAH88221.1}; HAMAP-Rule:MF_03038}; NBP; NBP 1; NBP 1 {ECO:0000255; NBP1; NBP35; NUBP1; nubp1 {ECO:0000312; nucleotide binding protein (e.coli MinD like); nucleotide binding protein 1; nucleotide binding protein 1 (E.coli MinD like); nucleotide binding protein 1 (MinD homolog, E. coli); nucleotide-binding protein 1; nucleotide-binding protein 1 {ECO:0000255
Gene Symbols NUBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUBP1. Recombinant protein control fragment (Product #RP-98659).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.