Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUBPL Monoclonal Antibody (OTI5C12), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503748S


Description
NUBPL Monoclonal antibody specifically detects NUBPL in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
NUBPL | |
Monoclonal | |
0.67 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8TB37 | |
Mouse | |
Affinity chromatography | |
RUO | |
80224 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI5C12 | |
Unconjugated | |
Nubpl | |
2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232 | |
Human recombinant protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction