Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUBPL Monoclonal Antibody (OTI5D5)
GREENER_CHOICE

Catalog No. PIMA525734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525734 Supplier Invitrogen™ Supplier No. MA525734
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NUBPL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUBPL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI5D5
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nubpl
Gene Accession No. Q8TB37
Gene Alias 2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232
Gene Symbols Nubpl
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-250 of NUBPL produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80224
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.