Learn More
NUBPL Mouse anti-Human, Clone: OTI5F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503729

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.Specifications
NUBPL | |
Monoclonal | |
Unconjugated | |
NUBPL | |
C14orf127, IND1, huInd1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80224 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI5F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8TB37 | |
NUBPL | |
Human recombit protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.