Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUBPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556083 Supplier Invitrogen™ Supplier No. PA556083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSRGLPKQKP IEGVKQVIVV ASGKGGVGKS TTAVNLALAL AANDSSKAIG LLDVDVYGPS VPKMMNLKGN PELSQSNLMR PLLNYGIACM SMGFLVEESE PVVWRG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

Required for the assembly of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I). May deliver of one or more Fe-S clusters to complex I subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUBPL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nubpl
Gene Accession No. Q8TB37
Gene Alias 2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232
Gene Symbols Nubpl
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUBPL. Recombinant protein control fragment (Product #RP-89006).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.