Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP BIOMEDICALS Nuc-Off Nucleases and Nucleic Acid Removal Spray

Catalog No. MP112461450 Shop All MP Biomedicals, Inc Products
Click to view available options
Volume (Metric):
450 mL
1800 mL

Non-toxic spray designed to remove nucleases and DNA.

Biomedicals Nuc-Off Nucleases and DNA Removal Spray is a ready-to-use solution for fast and cost-effective removal of RNase, DNase and DNA contamination.

This non-toxic spray-type solution can effectively remove nucleases and DNA from the surface of:

  • laboratory consumables
  • instruments
  • other utensils

creating a clean environment for RNA or DNA-related experiments.

It is strongly recommended to regularly clean your:

  • pipettors
  • bench surfaces
  • instrument exterior

Application Notes:

Nuc-Off Nucleases and DNA Removal Spray is intended for effective and safe elimination of Nucleases and DNA that may cause false positive or false negative experimental results.

Usage Statement:

Research Use Only (RUO)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Removal Agent
Volume (Metric) 450 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.