Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUCB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579767
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HEPA whole cell. IHC: mouse gaster tissue, rat gaster tissue.
NUCB2 was initially identified as calcium-binding protein EF hand motif-containing protein that could bind with Necdin, a growth suppressor expressed primarily in postmitotic neurons. NUC2 is a secreted protein that is cleaved into three major peptide products, Nesfatin-1, Nesfatin-2, and Nesfatin-3, of which Nesfatin-1 has been found to cause the suppression of food intake in a leptin-independent manner. Other studies have suggested that NUCB2/Nesfatin-1 may also play roles in energy homeostasis and closely related neuroendocrine functions.
Specifications
NUCB2 | |
Polyclonal | |
Unconjugated | |
NUCB2 | |
AI607786; Calnuc; DNA-binding protein NEFA; epididymis secretory protein Li 109; Gastric cancer antigen Zg4; HEL-S-109; LOW QUALITY PROTEIN: nucleobindin-2; NEFA; nesfatin 1; nesfatin-1; Nucb2; nucleobindin 2; nucleobindin-2; Nucleobindin-2/Nesfatin-1; nucleobindin-2-like protein; nucleobinding 2; p54; prepronefastin; prepronesfatin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53322, 59295 | |
-20°C | |
Lyophilized |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
P81117, Q9JI85 | |
NUCB2 | |
E. coli-derived mouse Nucleobindin 2 recombinant protein (Position: V25-L106). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction