Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NucGreen™ Dead 488 ReadyProbes™ Reagent (SYTOX™ Green)

Catalog No. R37109
Encompass_Preferred
Change view
Click to view available options
Quantity:
6 Bottles kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37109 6 Bottles kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37109 Supplier Invitrogen™ Supplier No. R37109
  • $370.00
    $332.65 / Each of 1

    Save $37.35

    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

NucGreen™ Dead 488 ReadyProbes™ Reagent

NucGreen Dead 488 ReadyProbes Reagent is a cell-impermeant stain that emits exceptionally bright, green fluorescence when bound to DNA. Cells that have lost plasma membrane integrity are stained within minutes, making this an extremely useful stain to estimate live/dead cell ratios and to measure cytotoxicity in kinetic live-cell assays. It is also suitable for staining nuclei in fixed cell preparations and tissue sections.

• Exceptionally bright green fluorescence upon binding to DNA
• Rapid staining of damaged, dead, or fixed cells without wash steps
• Ready-to-use liquid formulation in convenient dropper bottle—no need to dilute, weigh, or pipette
• Stability at room temperature—keep handy at your scope or cell

Cell imaging applications

The membrane-impermeable NucGreen Dead 488 reagent is a high-affinity DNA stain that easily stains cells with compromised cell membranes, yet does not enter living cells. It is therefore ideal for discrimination of live and dead cells. The fluorescence increases more than 500-fold on binding to dsDNA in cells with compromised cell membranes (dying or dead cells), in fixed cells, and tissue slices. The spectral properties, with excitation/emission at 504/523 nm when bound to DNA, are ideal for standard FITC filters.

Suggestions for use

• NucGreen Dead 488 reagent may be added directly to cells in full media or buffer solutions
• It easily stains nuclei of fixed cells and tissue slices
• In most cases, 2 drops/ml and an incubation time of 15 to 30 minutes will give bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained. In most cases, staining intensity increases with time if cells are not washed prior to imaging.
• NucGreen Dead 488 dye is excited with a maximum at 504 nm when bound to DNA, with an emission maximum at 523 nm. It is detected through standard GFP and FITC filters.

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-impermeant
Color Green
Content And Storage 6 x 2.5 mL dropper bottles

Store at ≤ 25°C
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Confocal Microscope, Floid Cell Imaging System, Fluorescence Microscope, Flow Cytometer
Product Type Nucleic Acid Stain
Dye Type Other Label(s) or Dye(s)
Form Liquid
Product Line NucGreen, ReadyProbes
Quantity 6 Bottles kit
Sub Cellular Localization Nucleus
Excitation Wavelength 504 nm
Emission Wavelength 523
Show More Show Less
Can I use the ReadyProbes reagents for flow cytometry?

This is not recommended. The ReadyProbes reagents were developed for imaging applications whereas the Ready Flow reagents were optimized for flow cytometry.

What is the difference between NucBlue Live reagents stain and NucGreen Dead reagents stain?

The NucBlue Live reagent stains the nuclei of all cells, while NucGreen Dead reagent stains only the nuclei of dead cells.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.