Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NUCKS1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074634AF532
Description
NUCKS1 Polyclonal antibody specifically detects NUCKS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NUCKS1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ21480, FLJ32016, FLJ38536, NUCKSnuclear ubiquitous casein and cyclin-dependent kinases substrate, nuclear casein kinase and cyclin-dependent kinase substrate 1, nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate, P1, potential LAG1 interactor | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate using the numbering given in entry NP_073568.2 (GeneID 64710). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64710 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction