Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NUCKS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526535
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526535 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526535 Supplier Thermo Scientific Supplier No. PA526535
Add to Cart
Add to Cart

NUCKS1 Polyclonal Antibody

NUCKS1 Polyclonal Antibody for Western Blot, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen NUCKS1
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Nucks1
Gene Accession No. Q9H1E3
Gene Alias 2700010L10Rik; 8430423A01Rik; AI647518; C78391; cyclin-dependent kinase substrate; JC7; Nucks; NUCKS1; nuclear casein kinase and cyclin dependent kinase substrate 1; nuclear casein kinase and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinases substrate; nuclear ubiquitous casein kinase 2; nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate; nuclear, casein kinase and cyclin-dependent kinase substrate; P1; potential LAG1 interactor
Gene Symbols Nucks1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 45-73 amino acids from the N-terminal region of human NUCKS1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.