Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUCKS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564016 Supplier Invitrogen™ Supplier No. PA564016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEDDDDSDYG SSKKKNKKMV KKSKPERKEK KMPKPRLKAT VTPSPVKGKG Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 100%.

This gene encodes a nuclear protein that is highly conserved in vertebrates. The conserved regions of the protein contain several consensus phosphorylation sites for casein kinase II and cyclin-dependent kinases, two putative nuclear localization signals, and a basic DNA-binding domain. It is phosphorylated in vivo by Cdk1 during mitosis of the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUCKS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nucks1
Gene Accession No. Q9H1E3
Gene Alias 2700010L10Rik; 8430423A01Rik; AI647518; C78391; cyclin-dependent kinase substrate; JC7; Nucks; NUCKS1; nuclear casein kinase and cyclin dependent kinase substrate 1; nuclear casein kinase and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinase substrate 1; nuclear ubiquitous casein and cyclin-dependent kinases substrate; nuclear ubiquitous casein kinase 2; nuclear ubiquitous casein kinase and cyclin-dependent kinase substrate; nuclear, casein kinase and cyclin-dependent kinase substrate; P1; potential LAG1 interactor
Gene Symbols Nucks1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NUCKS1. Recombinant protein control fragment (Product #RP-103854).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64710
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.