Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Antigen Antibody (235-1), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234525AF488
Description
Nuclear Antigen Monoclonal specifically detects Nuclear Antigen in Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Nuclear Antigen | |
Monoclonal | |
Alexa Fluor 488 | |
Nuclei, Nucleus | |
Nuclei of human myeloid leukemia biopsy cells | |
0.1 mL | |
Primary | |
Human, Chicken (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
235-1 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Intracellular) | |
Mouse | |
Protein A or G purified | |
RUO | |
This monoclonal antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. monoclonal antibody 235-1 recognizes an antigen associated with the nuclei in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in subcellular fractions. It produces a speckled pattern in normal and malignant cells and may be used to stain the nuclei of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction