Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Antigen Antibody (NM106), Alexa Fluor™ 350 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311613AF350
Description
Nuclear Antigen Monoclonal antibody specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Nuclear Antigen | |
Monoclonal | |
Alexa Fluor 350 | |
Nuclei, Nucleus | |
Nuclei of HL60 cells | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
NM106 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction