Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nuclear antigen Mouse anti-Human, Rat, Clone: NM106, Abnova™

Catalog No. 89034720 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-720 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-720 Supplier Abnova Corporation Supplier No. MAB14637
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against nuclear antigen.

  • High specificity
  • High reactivity

Specifications

Antigen Nuclear antigen
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone NM106
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against nuclear antigen.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunocytochemistry (0.25-0.5 ug/mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should b
Formulation In 10 mM PBS (0.05% BSA and 0.05% azide).
Host Species Mouse
Immunogen Nuclei of HL60 cells.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes an antigen associated with the nuclei in all cells.
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.