Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Antigen Rabbit anti-Human, Mouse, Rat, Clone: NM2984R, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP307303100UG
Description
Nuclear Antigen Monoclonal specifically detects Nuclear Antigen in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Nuclear Antigen | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry-Paraffin 0.25-0.5 μg/mL | |
Rabbit | |
Protein A or G purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry (Paraffin) | |
NM2984R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Nuclei of HL60 cells | |
100 μg | |
Primary | |
Store at 4°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction