Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Factor Erythroid Derived 2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310897100UL
Description
Nuclear Factor Erythroid Derived 2 Polyclonal specifically detects Nuclear Factor Erythroid Derived 2 in Mouse samples. It is validated for Western Blot.Specifications
Nuclear Factor Erythroid Derived 2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Leucine zipper protein NF-E2, NF-E2, nuclear factor (erythroid-derived 2), 45kD, nuclear factor (erythroid-derived 2), 45kDa, Nuclear factor, erythroid-derived 2 45 kDa subunit, p45, p45 NF-E2, transcription factor NF-E2 45 kDa subunit | |
The immunogen is a synthetic peptide directed towards the C-terminal region of MOUSE Nuclear Factor Erythroid Derived 2 (NP_032711). Peptide sequence ARGEADRTLEVMRQQLAELYHDIFQHLRDESGNSYSPEEYVLQQAADGAI | |
100 μg | |
Primary | |
Mouse | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
4778 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction