Learn More
Invitrogen™ Nuclear Matrix Protein p84 Monoclonal Antibody (301CT40.5.1)

Description
Nuclear Matrix Protein p84 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Nuclear Matrix Protein p84 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 301CT40.5.1 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Thoc1 |
| Gene Accession No. | Q96FV9 |
| Gene Alias | 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.