Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Matrix Protein p84 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596965
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596965 Supplier Invitrogen™ Supplier No. PA596965
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKKILMGNEE LTRLWNLCPD NMEACKSETR EHMPTLEEFF EEAIEQADPE NMVENEYKAV NNSNYGWRAL RLLARRSPHF FQPTNQQFKS LPEYLENMVI KLAKELPPPS EEIKTGEDED EEDNDALLKE NESPDVRRDK PVTGEQIEVF ANKLGEQWKI LAPYLEMKDS EIRQIECDSE DMKMRAKQLL VAWQDQEGVH ATPENLINAL NKSGLSDLAE SLTNDNETNS; Positive Samples: BT-474, MCF7, A-549, HeLa, HepG2; Cellular Location: Cytoplasm, Cytoplasm, Nucleus, Nucleus matrix, Nucleus speckle, nucleoplasm.

HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Matrix Protein p84
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Thoc1
Gene Accession No. P59924, Q8R3N6, Q96FV9
Gene Alias 3110002N20Rik; AW107452; HPR1; hTREX84; I79_014746; LRRGT00070; NMP-84; Nuclear matrix protein p84; P84; P84N5; THO complex 1; THO complex subunit 1; THO complex subunit 1-like protein; tho1; Thoc1
Gene Symbols Thoc1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 428-657 of human Nuclear Matrix Protein p84 (Nuclear Matrix Protein p84 (THOC1)) (NP_0051222).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225160, 291797, 9984
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.