Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Matrix Protein p84/THOC1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2448R

Description
HPR1 is part of the TREX (transcription/export) complex, which includes TEX1 (MIM 606929), THO2 (MIM 300395), ALY (MIM 604171), and UAP56 (MIM 142560).Specifications
Nuclear Matrix Protein p84/THOC1 | |
Polyclonal | |
Unconjugated | |
Thoc1 | |
3110002N20Rik; AW107452; BOS_22300; HPR1; hTREX84; I79_014746; LOC100350164; LRRGT00070; NMP-84; Nuclear matrix protein p84; p84; p84N5; THO complex 1; THO complex subunit 1; Tho1; THOC1 | |
Rabbit | |
Protein A | |
RUO | |
9984 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96FV9 | |
Thoc1 | |
KLH conjugated synthetic peptide derived from human HPR1/Nuclear Matrix Protein p84. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction