Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nuclear Membrane Marker Antibody (AE-5), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311308AF700
Description
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for ImmunofluorescenceSpecifications
Nuclear Membrane Marker | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Nuclei of myeloid leukemia biopsy cells | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunofluorescence | |
AE-5 | |
Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction