Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Membrane Monoclonal Antibody (IPO-38)
GREENER_CHOICE

Catalog No. 01670362
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-362 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-362 Supplier Invitrogen™ Supplier No. MA17628
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7628 detects Nuclear protein from human samples. MA1-7628 has been successfully used in ELISA procedures. The MA1-7628 immunogen is a proliferating Harvey cell line. Short term storage is at 4°C. For extended storage, aliquot and store at -80°C.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Nuclear Membrane
Applications ELISA, Western Blot
Classification Monoclonal
Clone IPO-38
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias Histone H1
Host Species Mouse
Immunogen Proliferating Harvey cell line.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.