Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Nucleic Acid Detection Blocking Buffer PROMO
Click to view available options
Quantity:
500 mL
Description
Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component. Non-specific binding of probe to membrane can be minimized by incubating the membrane in Blocking Buffer for 15 minutes.
Related Products
- Chemiluminescent Nucleic Acid Detection Module Kit (Cat. No. 89880)
Specifications
Specifications
Content And Storage | Store at 4°C. |
Buffer | Blocking Buffer |
Product Type | Nucleic Acid Detection Blocking Buffer |
Quantity | 500 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction