Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nucleobindin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592113
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592113 Supplier Invitrogen™ Supplier No. PA592113
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, HepG2, 293T, BxPC-3; Cellular Location: Cytoplasm, Golgi apparatus, Lumenal side, Peripheral membrane protein, cis-Golgi network membrane.

Nucleobindin 1 encodes a member of a small calcium-binding EF-hand protein family. The encoded protein is thought to have a key role in Golgi calcium homeostasis and Ca(2+)-regulated signal transduction events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleobindin 1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NUCB1
Gene Accession No. Q02818, Q02819, Q63083
Gene Alias B230337F23Rik; Bone 63 kDa calcium-binding protein; C77483; CALNUC; MTEST82; NUC; Nucb; Nucb1; nucleobindin 1; nucleobindin-1
Gene Symbols NUCB1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18220, 4924, 84595
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.