Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleocapsid Antibody (1051436), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB11289Z
Description
Nucleocapsid Monoclonal antibody specifically detects Nucleocapsid in HCoV-OC43 samples. It is validated for Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
Nucleocapsid | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Sf-21 (baculovirus)-derived hcov-oc43 Nucleocapsid, Met1-Ile448, Accession # YP_009555245.1 | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry, Immunocytochemistry | |
1051436 | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
HCoV-OC43 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction