Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleocapsid Protein Rabbit, anti-SARS-CoV-2, Unconjugated, MP Biomedicals™

Catalog No. MP08L100001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100001 Supplier MP Biomedicals, Inc Supplier No. 08L100001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SARS-CoV-2 Nucleoprotein Rabbit Monoclonal Antibody has been experimentally validated in WB, ELISA, FCM, IHC, IF, IP assays.

  • Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP) and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M
  • Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication
  • Offers high specificity and potency against SARS-CoV-2 antigens in immunological and biochemical assays and consistency between lots
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleocapsid Protein
Applications ELISA, Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot: 1:1000 to 1:5000; ELISA: 1:1000 to 1:5000; Flow Cytometry: 1:50 to 1:200; Immunohistochemistry: 1:50 to 1:200; Immunofluorescence: 1:50 to 1:200; Immunoprecipitation: 1:50 to 1:200
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol, pH7.3 to 7.4.
Gene Alias Nucleocapsid protein; NP; Nucleoprotein; Protein N
Host Species Rabbit
Immunogen Full-length SARS-CoV-2 nucleocapsid protein
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.