Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleolin Antibody (364-5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247861MFV450
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Nucleolin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
364-5 | |
Flow Cytometry | |
C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
lysate of SU-DHL-1 Nuclei (Uniprot: P19338) | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
4691 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction