Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleolin Antibody (364-5 + NCL/902) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2478620.2MG
Description
Nucleolin Monoclonal antibody specifically detects Antigen in Bovine (Negative), Human, Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.Specifications
Nucleolin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot : 0.5 - 1.0 ug/ml, Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
Mouse | |
76 kDa | |
0.2 mg | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
4691 | |
Human, Bovine (Negative), Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
364-5 + NCL/902 | |
Unconjugated | |
PBS with No Preservative | |
NCL | |
Lysate of SU-DHL-1 Nuclei (364-5); Recombinant human NCL protein (NCL/902) | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of ∼76kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly. This MAb can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces a speckled pattern in the nuclei of cells of normal and malignant cells and may be used to stain the nucleoli of cells in fixed or frozen tissue sections. It can be used with paraformaldehyde fixed frozen tissue or cell preparations and formalin fixed, paraffin-embedded tissue sections. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction