Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleolin Antibody (NCL/7338), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314014AF750
This item is not returnable.
View return policy
Description
Nucleolin Monoclonal antibody specifically detects Nucleolin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Nucleolin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| C23, FLJ45706, FLJ59041, nucleolin, Protein C23 | |
| Recombinant full-length human Nucleolin protein (Uniprot: P19338) | |
| 0.1 mL | |
| Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA Replication Transcription Translation and Splicing, Stem Cell Markers | |
| 4691 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| NCL/7338 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction