Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Antibody, Novus Biologicals™
SDP

Catalog No. p-5087660 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001920SS 0.025 mL
NB1001920 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001920SS Supplier Novus Biologicals Supplier No. NB1001920SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Nucleolin Polyclonal specifically detects Nucleolin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen Nucleolin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, Immunohistochemistry 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:3000-1:10000, Immunoprecipitation 1-3ug, Immunohistochemistry-Paraffin 1:5000-1:10000, Knockdown Validated
Formulation Whole antisera with No Preservative
Gene Accession No. P19338
Gene Alias C23, FLJ45706, FLJ59041, nucleolin, Protein C23
Gene Symbols NCL
Host Species Rabbit
Immunogen A 15 residue peptide designed to the C-terminus of human Nucleolin [UniProt# P19338]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4691
Target Species Human, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.