Learn More
Nucleolin Monoclonal Antibody (3G4B20), Active Motif™

Description
Specifications
Specifications
| Antigen | Nucleolin |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 3G4B20 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | antibody; pH |
| Gene | NCL |
| Gene Accession No. | P09405, P19338 |
| Gene Alias | B530004O11Rik; BOS_2214; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; NCL; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.