Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Monoclonal Antibody (3G4B20), Active Motif™
SDP

Catalog No. 501994043
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4043 10 μL
50-199-4042 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4043 Supplier Active Motif Supplier No. 39542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.

Specifications

Antigen Nucleolin
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3G4B20
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation antibody; pH
Gene NCL
Gene Accession No. P09405, P19338
Gene Alias B530004O11Rik; BOS_2214; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; NCL; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23
Gene Symbols NCL
Host Species Mouse
Immunogen This Nucleolin antibody was raised against a salt-extracted nuclear matrix preparation from MDCk cells.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17975, 4691
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.