Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nucleolin Monoclonal Antibody (4i51)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA125778
Description
MA1-25778 will not react with mouse or rat. Suggested positive controls for this product are HeLa nuclear extract/HeLa cells.
Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.
Specifications
Nucleolin | |
Monoclonal | |
Conc. Not Determined | |
Tissue culture supernatant with 30% glycerol and 0.05% sodium azide | |
P19338 | |
NCL | |
Nuclear matrix extract from human MDCK cells. | |
RUO | |
4691, 608445 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
4i51 | |
Unconjugated | |
NCL | |
B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23 | |
Mouse | |
25 μL | |
Primary | |
Human, Canine, Monkey | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction