Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nucleolin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519508
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519508 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519508 Supplier Invitrogen™ Supplier No. PA519508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat mediated antigen retrieval recommended prior to tissue staining. Store antibody at 4ºC for 1-2 weeks. For long-term storage, store at -20ºC.

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleolin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene NCL
Gene Accession No. P09405, P13383, P19338
Gene Alias B530004O11Rik; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; Ncl; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23
Gene Symbols NCL
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human Nucleolin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17975, 25135, 4691
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.