Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleolin Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™
SDP

Catalog No. 501993762
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3762 10 μg
50-199-3761 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3762 Supplier Active Motif Supplier No. 91232
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Nucleolin (NCL), a eukaryotic nucleolar phosphoprotein, is involved in the synthesis and maturation of ribosomes. It is located mainly in dense fibrillar regions of the nucleolus. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. The intron 11 of the NCL gene encodes a small nucleolar RNA, termed U20.

Specifications

Antigen Nucleolin
Applications Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5
Gene NCL
Gene Accession No. P19338
Gene Alias B530004O11Rik; BOS_2214; C23; D0Nds28; D1Nds28; FLJ45706; FLJ59041; NCL; Nuc; Nucl; nucl antibody; Nucleolin; nucleolin-like protein; Protein C23
Gene Symbols NCL
Host Species Mouse
Immunogen This antibody was raised against a salt-extracted nuclear matrix preparation from MDCk (canine kidney) cells.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4691, 608445
Target Species Canine, Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.