Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleoporin 107 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299074JF646
Description
Nucleoporin 107 Polyclonal specifically detects Nucleoporin 107 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Nucleoporin 107 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Nucleoporin 107. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
57122 | |
Store at 4°C in the dark. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction