Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleoporin 107 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176927MFV500
Description
Nucleoporin 107 Polyclonal antibody specifically detects Nucleoporin 107 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Nucleoporin 107 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
nuclear pore complex protein Nup107, nucleoporin 107kDa, NUP84Nucleoporin Nup107,107 kDa nucleoporin | |
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human NUP107. The immunogen is located within amino acids 820 - 870 of NUP107. Amino Acid Squence: DVREDAKEDHERTHQ | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
57122 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction