Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleoprotein TPR Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5755R
Description
Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.Specifications
| Nucleoprotein TPR | |
| Polyclonal | |
| Unconjugated | |
| TPR | |
| 2610029M07Rik; BOS_16180; C77892; LOC100346739; Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein Tpr; Nucleoprotein TPR; TPR; translocated promoter region (to activated MET oncogene); Translocated promoter region and nuclear basket protein; Translocated promoter region protein; translocated promoter region, nuclear basket protein; tumor potentiating region | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7175 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| P12270 | |
| TPR | |
| KLH conjugated synthetic peptide derived from human Nucleoprotein TPR. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction