Learn More
Nucleoredoxin/NXN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304985AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Zebrafish, X. laevis and Bovine
Functions as a redox-dependent negative regulator of the Wnt signaling pathway, possibly by preventing ubiquitination of DVL3 by the BCR(KLHL12) complex. May also function as a transcriptional regulator act as a regulator of protein phosphatase 2A (PP2A).Specifications
Nucleoredoxin/NXN | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P97346, Q6DKJ4 | |
NXN | |
Between 385 and 435 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NXN | |
Nucleoredoxin, NXN, NRX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18230, 64359 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.