Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nucleoredoxin/NXN Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501569200
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9200 Supplier Bethyl Laboratories, Inc Supplier No. A304985A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Zebrafish, X. laevis and Bovine

Functions as a redox-dependent negative regulator of the Wnt signaling pathway, possibly by preventing ubiquitination of DVL3 by the BCR(KLHL12) complex. May also function as a transcriptional regulator act as a regulator of protein phosphatase 2A (PP2A).

Specifications

Antigen Nucleoredoxin/NXN
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene NXN
Gene Accession No. P97346, Q6DKJ4
Gene Alias Nucleoredoxin, NXN, NRX
Gene Symbols NXN
Host Species Rabbit
Immunogen Between 385 and 435
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18230, 64359
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.