Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleostemin Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298743AF700
Description
Nucleostemin Polyclonal specifically detects Nucleostemin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Nucleostemin | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Alexa Fluor 700 | |
C77032, E2IG3NNP47, estradiol-induced nucleotide binding protein, guanine nucleotide binding protein-like 3 (nucleolar), guanine nucleotide-binding protein-like 3, MGC800, Novel nucleolar protein 47, NSE2-induced gene 3 protein, Nucleolar GTP-binding protein 3, nucleostemin | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Nucleostemin. | |
0.1 mL | |
Apoptosis, Cellular Markers, Core ESC Like Genes, Epigenetics, mTOR Pathway, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
26354 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction