Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nucleostemin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143898
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143898 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143898 Supplier Invitrogen™ Supplier No. PA5143898
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human K562 whole cell, human PC-3 whole cell, human Jurkat whole cell, rat C6 whole cell, mouse NIH/3T3 whole cell. IHC: human appendiceal adenocarcinoma tissue, human breast cancer tissue, human colonic adenocarcinoma tissue, human gall bladder adenosquamous carcinoma tissue. ICC/IF: HEP3B cell. Flow: Hela cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Arylsulfatase F, also known as ARSF, is a 590 amino acid secretory protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Arylsulfatase F uses calcium as a cofactor to catalyze reactions that are important in maintaining correct bone composition. The activity of Arylsulfatase F, unlike that of other family members, such as Arylsulfatase E, is not inhibited by warfarin. The gene encoding Arylsulfatase F maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleostemin
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GNL3
Gene Accession No. Q811S9, Q8CI11, Q9BVP2
Gene Alias C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin
Gene Symbols GNL3
Host Species Rabbit
Immunogen E.coli-derived human Nucleostemin/GNL3 recombinant protein (Position: R45-V549).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26354, 290556, 30877
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.