Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nucleostemin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115305
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115305 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115305 Supplier Invitrogen™ Supplier No. PA5115305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNL3.

Arylsulfatase F, also known as ARSF, is a 590 amino acid secretory protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Arylsulfatase F uses calcium as a cofactor to catalyze reactions that are important in maintaining correct bone composition. The activity of Arylsulfatase F, unlike that of other family members, such as Arylsulfatase E, is not inhibited by warfarin. The gene encoding Arylsulfatase F maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleostemin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GNL3
Gene Accession No. Q811S9, Q8CI11, Q9BVP2
Gene Alias C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin
Gene Symbols GNL3
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNL3(Accession Q9BVP2), corresponding to amino acid residues K23-E73.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26354, 290556, 30877
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.