Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleostemin Polyclonal Antibody
Nucleostemin Polyclonal Antibody
Supplier: Thermo Scientific PA592992
Description
Nucleostemin Polyclonal Antibody for Western Blot, ICC/IF, IP

Specifications
Nucleostemin | |
Polyclonal | |
Unconjugated | |
GNL3 | |
C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26354 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BVP2 | |
GNL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human GNL3 (NP_9965621). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction