Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nucleostemin Recombinant Rabbit Monoclonal Antibody (JG37-82)
GREENER_CHOICE

Catalog No. PIMA542483
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542483 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542483 Supplier Invitrogen™ Supplier No. MA542483
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: A549, HepG2, human lung cancer tissue, human breast cancer tissue, human skin tissue. Subcellular Location: Nucleus.

Arylsulfatase F, also known as ARSF, is a 590 amino acid secretory protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Arylsulfatase F uses calcium as a cofactor to catalyze reactions that are important in maintaining correct bone composition. The activity of Arylsulfatase F, unlike that of other family members, such as Arylsulfatase E, is not inhibited by warfarin. The gene encoding Arylsulfatase F maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nucleostemin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JG37-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene GNL3
Gene Accession No. Q9BVP2
Gene Alias C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin
Gene Symbols GNL3
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal Human Nucleostemin .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26354
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.