Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleotide binding protein like Antibody (OTI5D5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273094FR
Description
Nucleotide binding protein like Monoclonal specifically detects Nucleotide binding protein like in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Nucleotide binding protein like | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
NUBPL | |
Recombinant protein fragment corresponding to amino acids 1-250 of human Nucleotide binding protein like (NP_079428) produced in HEK293T cells. | |
0.1 mL | |
80224.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
C14orf127, FLJ12660, huInd1IND1 homolog, IND1chromosome 14 open reading frame 127, iron-sulfur protein NUBPL, iron-sulfur protein required for NADH dehydrogenase, nucleotide binding protein-like, Nucleotide-binding protein-like | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction