Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nucleotide binding protein like Antibody (OTI5D5), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273094MFV610
Description
Nucleotide binding protein like Monoclonal antibody specifically detects Nucleotide binding protein like in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Nucleotide binding protein like | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
80224 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI5D5 | |
Flow Cytometry | |
C14orf127, FLJ12660, huInd1IND1 homolog, IND1chromosome 14 open reading frame 127, iron-sulfur protein NUBPL, iron-sulfur protein required for NADH dehydrogenase, nucleotide binding protein-like, Nucleotide-binding protein-like | |
Human recombinant protein fragment corresponding to amino acids 1-250 of human NUBPL(NP_079428) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction