Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NUDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530973 Supplier Invitrogen™ Supplier No. PA530973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, BCL-1. Predicted reactivity: Mouse (90%), Rat (92%), Pig (92%), Chimpanzee (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NUDC
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene NUDC
Gene Accession No. O35685, Q9Y266
Gene Alias c15; clone 15; H M NPD011; HNUDC; MNUDC; MNUDC protein; NPD011; nuclear distribution C homolog; nuclear distribution C, dynein complex regulator; nuclear distribution gene C homolog; nuclear distribution gene C homolog (Aspergillus); nuclear distribution protein C homolog; Nuclear migration protein nudC; Nudc; nudC nuclear distribution protein; OTTHUMP00000004405; SIG-92; Silg92; silica-induced gene 92 protein
Gene Symbols NUDC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NUDC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10726, 18221
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.