Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NUDC Recombinant Rabbit Monoclonal Antibody (JE50-74)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA534878
Description
Positive Control: K562 cell lysates, PC-3M cell lysates, U937 cell lysates, rat brain tissue, human tonsil tissue, human colon carcinoma tissue, human gastric carcinoma tissue, mouse brain tissue, F9.
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
Specifications
NUDC | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y266 | |
NUDC | |
Recombinant protein within Human NUDC aa 207-331. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
JE50-74 | |
Unconjugated | |
NUDC | |
c15; clone 15; H M NPD011; HNUDC; MNUDC; MNUDC protein; NPD011; nuclear distribution C homolog; nuclear distribution C, dynein complex regulator; nuclear distribution gene C homolog; nuclear distribution gene C homolog (Aspergillus); nuclear distribution protein C homolog; Nuclear migration protein nudC; Nudc; nudC nuclear distribution protein; OTTHUMP00000004405; SIG-92; Silg92; silica-induced gene 92 protein | |
Rabbit | |
Protein A | |
RUO | |
10726 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction